Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159387.1 | NP_001152859.1 | 9 | 1022 | ||
NM_001159388.1 | NP_001152860.1 | 9 | 1035 | |||
NM_153446.2 | NP_703147.2 | 9 | 1237 | |||
XM_011524314.2 | XP_011522616.1 | 8 | 1202 | |||
XM_017024173.1 | XP_016879662.1 | 9 | 1084 | |||
GenBank mRNA | AF510036.1 | - | 9 | 1237 | ||
AJ517770.1 | - | 9 | 1237 | |||
AJ517771.1 | - | 9 | 1019 | |||
AK302876.1 | - | 9 | 1035 | |||
BC113675.1 | - | 9 | 1231 | 8322727 | ||
BC113677.1 | - | 9 | 1231 | 8322729 | ||
S83275.1 | - | 3 | 299 |