Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152345.4 | NP_689558.4 | 8, 9 | 1018 | ||
XM_005257920.4 | XP_005257977.1 | 8, 9 | 951 | |||
XM_011524319.2 | XP_011522621.1 | 8, 9 | 1188 | |||
XM_011524320.2 | XP_011522622.1 | 8, 9 | 950 | |||
XM_011524321.2 | XP_011522623.1 | 8, 9 | 950 | |||
XM_017024174.1 | XP_016879663.1 | 8, 9 | 885 | |||
XM_017024175.1 | XP_016879664.1 | 8, 9 | 887 | |||
XM_017024176.1 | XP_016879665.1 | 8, 9 | 817 | |||
GenBank mRNA | AK092673.1 | - | 8, 9 | 835 | ||
BC032554.1 | - | 8, 9 | 1065 | 5557374 | ||
BC062597.1 | - | 7, 8 | 868 |