Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 4 | 735 | ||
NM_001321364.1 | NP_001308293.1 | 4 | 533 | |||
NM_001321365.1 | NP_001308294.1 | 4 | 702 | |||
NM_152346.2 | NP_689559.1 | 4 | 533 | |||
XM_017024177.1 | XP_016879666.1 | 4 | 710 | |||
XM_017024178.1 | XP_016879667.1 | 4 | 743 | |||
XM_017024179.1 | XP_016879668.1 | 3 | 642 | |||
GenBank mRNA | AB120364.1 | - | 4 | 701 | ||
AF289592.1 | - | 4 | 578 | |||
AK075068.1 | - | 2 | 73 | |||
AK075173.1 | - | 2 | 127 | |||
BC027923.1 | - | 4 | 483 | 5309751 | ||
BC071859.1 | - | 4 | 533 | 6572285 |