Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 15 | 1956 | ||
NM_001284499.1 | NP_001271428.1 | 10 | 1397 | |||
NM_001321364.1 | NP_001308293.1 | 15 | 1754 | |||
NM_001321365.1 | NP_001308294.1 | 15 | 1923 | |||
NM_152346.2 | NP_689559.1 | 14 | 1742 | |||
XM_017024177.1 | XP_016879666.1 | 14 | 1919 | |||
XM_017024178.1 | XP_016879667.1 | 14 | 1952 | |||
XM_017024179.1 | XP_016879668.1 | 13 | 1851 | |||
GenBank mRNA | AB120364.1 | - | 15 | 1922 | ||
AK074428.1 | - | 4 | 273 | |||
AK075068.1 | - | 12 | 1282 | |||
AK075173.1 | - | 12 | 1336 | |||
AK075196.1 | - | 8 | 1023 | |||
AK301127.1 | - | 10 | 1280 | |||
BC019562.1 | - | 8 | 1017 | |||
BC027923.1 | - | 14 | 1692 | 5309751 |