Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 6 | 876 | ||
NM_001284499.1 | NP_001271428.1 | 2 | 329 | |||
NM_001321364.1 | NP_001308293.1 | 6 | 674 | |||
NM_001321365.1 | NP_001308294.1 | 6 | 843 | |||
NM_152346.2 | NP_689559.1 | 6 | 674 | |||
XM_017024177.1 | XP_016879666.1 | 6 | 851 | |||
XM_017024178.1 | XP_016879667.1 | 6 | 884 | |||
XM_017024179.1 | XP_016879668.1 | 5 | 783 | |||
GenBank mRNA | AB120364.1 | - | 6 | 842 | ||
AF289592.1 | - | 6 | 719 | |||
AK075068.1 | - | 4 | 214 | |||
AK075173.1 | - | 4 | 268 | |||
AK301127.1 | - | 2 | 212 | |||
BC027923.1 | - | 6 | 624 | 5309751 | ||
BC071859.1 | - | 6 | 674 | 6572285 | ||
DC381982.1 | - | 2 | 330 |