Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254755.1 | NP_001241684.1 | 2 | 233 | ||
NM_001254756.1 | NP_001241685.1 | 2 | 236 | |||
NM_144611.3 | NP_653212.1 | 2 | 928 | |||
XM_006721447.2 | XP_006721510.2 | 2 | 2065 | |||
XM_011523649.2 | XP_011521951.1 | 2 | 316 | |||
XM_017024180.1 | XP_016879669.1 | 2 | 319 | |||
XM_017024181.1 | XP_016879670.1 | 2 | 208 | |||
XM_017024182.1 | XP_016879671.1 | 2 | 2370 | |||
XM_017024183.1 | XP_016879672.1 | 2 | 622 | |||
GenBank mRNA | AA479992.1 | - | 1 | 36 | ||
AK172844.1 | - | 2 | 394 | |||
AK313088.1 | - | 2 | 399 | |||
BC020263.1 | - | 2 | 380 | 4877960 | ||
BC051697.1 | - | 2 | 840 | 5190840 | ||
CB112805.1 | - | 2 | 236 | |||
CB129650.1 | - | 2 | 203 | |||
CK821986.1 | - | 1 | 100 | |||
EB386356.1 | - | 2 | 233 |