Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254755.1 | NP_001241684.1 | 3, 4 | 477 | ||
NM_001254756.1 | NP_001241685.1 | 3, 4 | 480 | |||
NM_144611.3 | NP_653212.1 | 3, 4 | 1172 | |||
XM_006721447.2 | XP_006721510.2 | 3, 4 | 2309 | |||
XM_011523649.2 | XP_011521951.1 | 3, 4 | 560 | |||
XM_017024180.1 | XP_016879669.1 | 3, 4 | 563 | |||
XM_017024181.1 | XP_016879670.1 | 3, 4 | 452 | |||
XM_017024182.1 | XP_016879671.1 | 3, 4 | 2614 | |||
GenBank mRNA | AA479992.1 | - | 2, 3 | 279 | ||
AK172844.1 | - | 3, 4 | 638 | |||
AK313088.1 | - | 3, 4 | 643 | |||
BC020263.1 | - | 3, 4 | 624 | 4877960 | ||
BC051697.1 | - | 3, 4 | 1084 | 5190840 | ||
BX327481.2 | - | 2, 3 | 263 | |||
CB112805.1 | - | 3, 4 | 480 | |||
CB129650.1 | - | 3, 4 | 447 | |||
CK821986.1 | - | 2, 3 | 344 |