Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145255.3 | NP_660298.2 | 3 | 308 | ||
NM_148887.2 | NP_683685.1 | 4 | 406 | |||
NR_037575.1 | 3 | 457 | ||||
GenBank mRNA | AK127167.1 | - | 3 | 1183 | ||
AK293624.1 | - | 3 | 446 | |||
AL558270.3 | - | 4 | 371 | |||
AL701268.1 | - | 3 | 305 | |||
BC015904.1 | - | 3 | 279 | 3919892 | ||
BC039852.1 | - | 3 | 287 | |||
BC052601.1 | - | 3 | 297 | 6085551 | ||
DA137043.1 | - | 3 | 308 |