Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163673.1 | NP_001157145.1 | 4 | 498 | ||
NM_001163809.1 | NP_001157281.1 | 4 | 4183 | |||
NM_001163811.1 | NP_001157283.1 | 4 | 634 | |||
NM_152348.3 | NP_689561.2 | 5 | 1135 | |||
XM_011523651.2 | XP_011521953.1 | 5 | 1120 | |||
XM_017024184.1 | XP_016879673.1 | 4 | 4206 | |||
GenBank mRNA | AK074111.1 | - | 4 | 1437 | ||
AK091136.1 | - | 5 | 1119 | |||
AK122972.1 | - | 4 | 1445 | |||
AK123896.1 | - | 4 | 625 | |||
AK127946.1 | - | 4 | 634 | |||
AK298567.1 | - | 4 | 498 | |||
AL834379.1 | - | 4 | 959 | |||
BC092513.1 | - | 4 | 910 | 3092839 | ||
BC114519.1 | - | 3 | 393 | 40036610 | ||
BC114568.1 | - | 4 | 460 | 40036619 |