Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 8 | 1039 | ||
NM_001271845.1 | NP_001258774.1 | 8 | 1078 | |||
NM_019020.3 | NP_061893.2 | 12 | 2194 | |||
XM_005257049.2 | XP_005257106.1 | 12 | 2205 | |||
XM_005257050.3 | XP_005257107.1 | 12 | 2208 | |||
XM_006721694.3 | XP_006721757.1 | 12 | 2211 | |||
XM_011524322.2 | XP_011522624.1 | 12 | 2169 | |||
XM_011524323.2 | XP_011522625.2 | 10 | 2119 | |||
XM_011524324.2 | XP_011522626.1 | 8 | 1663 | |||
XM_017024186.1 | XP_016879675.1 | 10 | 1946 | |||
XM_017024187.1 | XP_016879676.1 | 11 | 2186 | |||
XM_017024188.1 | XP_016879677.1 | 10 | 1326 | |||
GenBank mRNA | AB449902.1 | - | 11 | 2068 | ||
BC028290.1 | - | 12 | 2184 | 4480654 | ||
BC036947.1 | - | 8 | 1083 | 5392579 | ||
CR936670.1 | - | 8 | 1044 |