Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFMID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010982.4 | NP_001010982.2 | 6 | 499 | ||
NM_001145526.2 | NP_001138998.1 | 6 | 499 | |||
XM_011524326.2 | XP_011522628.1 | 5 | 431 | |||
XM_011524327.2 | XP_011522629.1 | 7 | 667 | |||
XM_011524328.2 | XP_011522630.1 | 7 | 639 | |||
XM_011524329.2 | XP_011522631.1 | 7 | 681 | |||
XM_011524330.2 | XP_011522632.1 | 7 | 685 | |||
XM_011524331.2 | XP_011522633.1 | 6 | 581 | |||
XM_011524332.2 | XP_011522634.1 | 6 | 553 | |||
XM_011524333.2 | XP_011522635.1 | 6 | 458 | |||
XM_011524336.2 | XP_011522638.1 | 6 | 458 | |||
XM_011524337.2 | XP_011522639.1 | 5 | 414 | |||
GenBank mRNA | BC132824.1 | - | 6 | 499 | 40146846 | |
BX648442.1 | - | 6 | 455 |