Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152219.3 | NP_689343.3 | 1 | 1152 | ||
GenBank mRNA | AK125254.1 | - | 1 | 1152 |