Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000795.3 | NP_000786.1 | 3 | 532 | ||
NM_016574.3 | NP_057658.2 | 3 | 532 | |||
XM_017017296.1 | XP_016872785.1 | 3 | 514 | |||
GenBank mRNA | AB451480.1 | - | 2 | 297 | ||
AF176812.1 | - | 2 | 306 | |||
AK308777.1 | - | 3 | 362 | |||
BC021195.2 | - | 3 | 392 | 3939741 | ||
BF437994.1 | - | 3 | 364 | |||
M29066.1 | - | 2 | 330 | |||
M30625.1 | - | 3 | 633 | |||
S62137.1 | - | 2 | 614 | |||
S69899.1 | - | 3 | 396 | |||
X51362.1 | - | 3 | 462 | |||
X51645.1 | - | 2 | 325 |
Set Membership: |
G-protein coupled receptor |