Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRC75A-AS1 SIRNA Assays ›
Gene Symbol:
LRRC75A-AS1
Gene Name:
LRRC75A antisense RNA 1
Gene Aliases:
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
Chromosome Location:
Chr.17: 16438987 - 16470648 on Build GRCh38
UniGene ID:
Hs.368934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42866
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRC75A-AS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027158.1
3
174
NR_027159.1
2
440
NR_027160.1
3
563
NR_027161.1
3
616
NR_027162.1
3
616
NR_027163.1
4
350
NR_027164.1
4
297
NR_027165.1
4
218
NR_027166.1
3
174
NR_027167.1
4
297
NR_027168.2
4
266
NR_027169.1
4
350
NR_027170.1
4
218
NR_027171.1
2
440
NR_027172.2
3
143
NR_027173.1
3
174
NR_027174.1
3
174
NR_027175.1
3
174
NR_027176.1
3
174
NR_027177.1
3
174
NR_027178.1
3
174
NR_027179.1
3
174
NR_027667.1
3
484
NR_045021.1
3
153
NR_045022.1
4
292
NR_045023.1
3
174
NR_045024.1
2
440
NR_045025.1
2
86
NR_045026.1
4
350
NR_045027.1
2
440
NR_045028.1
3
484
NR_045029.1
3
248
GenBank mRNA
AK098643.1
-
3
124
AK307982.1
-
3
566
AK308482.1
-
3
124
AK309961.1
-
4
248
AK311247.1
-
3
730
BC012396.1
-
2
94
BC019833.1
-
2
100
BC027986.1
-
3
434
4940349
BC040159.1
-
3
413
5539041
BI333700.1
-
3
245
BP353162.1
-
4
300
BP377665.1
-
2
375
BP870476.1
-
4
168
BQ070126.1
-
2
178
BQ954204.1
-
3
540
BU165462.1
-
2
367
BU506593.1
-
281
BU599615.1
-
4
304
BX538291.1
-
4
169
CB150672.1
-
4
247
CD050014.1
-
3
147
CF126952.1
-
2
356
CN297624.1
-
3
143
CR989654.1
-
3
227
DA131201.1
-
3
124
DA193585.1
-
3
309
DA383448.1
-
3
124
DA440445.1
-
4
243
DA485648.1
-
3
125
DA494627.1
-
2
391
DA769571.1
-
3
124
DA793334.1
-
2
40
DA802938.1
-
3
124
DA857697.1
-
3
124
DA867958.1
-
4
302
DB258221.1
-
2
391
DW409012.1
-
3
141
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0