Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042697.1 | NP_001036162.1 | 4 | 343 | ||
NM_001042698.1 | NP_001036163.1 | 4 | 343 | |||
GenBank mRNA | AK093384.1 | - | 2 | 92 | ||
BC018807.1 | - | 4 | 314 | |||
BC044842.1 | - | 4 | 301 | |||
BC066344.1 | - | 4 | 335 | |||
BC089386.1 | - | 5 | 427 | |||
BC094855.1 | - | 6 | 625 | |||
BU075283.1 | - | 4 | 346 | |||
DQ530600.1 | - | 4 | 245 |