Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144900.1 | NP_001138372.1 | 4 | 2353 | ||
NM_139162.3 | NP_631901.2 | 4 | 2427 | |||
NM_148886.1 | NP_683684.2 | 4 | 2275 | |||
XM_017024190.1 | XP_016879679.1 | 4 | 2772 | |||
GenBank mRNA | AF467443.1 | - | 4 | 2275 | ||
AK056165.1 | - | 4 | 2406 | |||
AK128310.1 | - | 4 | 2256 | |||
BC014973.1 | - | 4 | 2275 | 4859126 | ||
BC035292.1 | - | 1 | 775 |