Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042461.2 | NP_001035926.1 | 2 | 319 | ||
NM_001042462.1 | NP_001035927.1 | 2 | 171 | |||
NM_174894.2 | NP_777554.1 | 2 | 342 | |||
GenBank mRNA | AK292001.1 | - | 2 | 149 | ||
BC042161.1 | - | 2 | 288 | 4869377 | ||
BG528980.1 | - | 2 | 172 | |||
BI196928.1 | - | 2 | 262 | |||
BM841051.1 | - | 2 | 159 |