Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271830.1 | NP_001258759.1 | 3, 4 | 305 | ||
NM_174895.2 | NP_777555.1 | 3, 4 | 587 | |||
XM_006722639.2 | XP_006722702.1 | 3, 4 | 807 | |||
XM_017026249.1 | XP_016881738.1 | 5, 6 | 1616 | |||
XM_017026250.1 | XP_016881739.1 | 5, 6 | 1475 | |||
XM_017026251.1 | XP_016881740.1 | 4, 5 | 1431 | |||
XM_017026252.1 | XP_016881741.1 | 3, 4 | 948 | |||
GenBank mRNA | AK130855.1 | - | 3, 4 | 539 | ||
BC025387.1 | - | 3, 4 | 592 | 4824135 | ||
BC038715.1 | - | 3, 4 | 523 | 5268414 | ||
BX281028.1 | - | 3, 4 | 264 | |||
BX647746.1 | - | 3, 4 | 335 |