Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282349.1 | NP_001269278.1 | 5 | 1789 | ||
NM_001282350.1 | NP_001269279.1 | 4 | 1756 | |||
NM_130771.4 | NP_570127.3 | 7 | 1287 | |||
NM_133168.4 | NP_573398.2 | 5 | 1242 | |||
NM_133169.4 | NP_573399.2 | 6 | 1275 | |||
NM_206818.2 | NP_996554.2 | 6 | 1801 | |||
GenBank mRNA | AF251702.1 | - | 5 | 1129 | ||
AF391162.1 | - | 5 | 1090 | |||
AF391163.1 | - | 6 | 1095 | |||
AF391164.1 | - | 4 | 1088 | |||
AF474152.1 | - | 4 | 1635 | |||
AF474153.1 | - | 3 | 1564 | |||
AK091111.1 | - | 4 | 1588 | |||
AK130199.1 | - | 5 | 1633 | |||
BC035023.2 | - | 5 | 1789 | 4825982 |