Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175871.3 | NP_787067.2 | 1 | 203 | ||
GenBank mRNA | AK092438.1 | - | 1 | 203 | ||
BC068071.1 | - | 1 | 225 | 5480023 | ||
BC119677.2 | - | 1 | 199 | 40116265 |