Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CPT1C SIRNA Assays ›
Gene Symbol:
CPT1C
Gene Name:
carnitine palmitoyltransferase 1C
Gene Aliases:
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location:
Chr.19: 49690672 - 49713731 on Build GRCh38
UniGene ID:
Hs.112195
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42960
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CPT1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136052.2
NP_001129524.1
13
1655
NM_001199752.2
NP_001186681.1
13
1752
NM_001199753.1
NP_001186682.1
13
1688
NM_152359.2
NP_689572.1
12
1619
NR_108072.1
14
1850
XM_005258505.2
XP_005258562.1
13
1725
XM_005258506.4
XP_005258563.1
12
1641
XM_006723009.3
XP_006723072.1
10
1457
XM_011526438.2
XP_011524740.1
13
1781
XM_011526439.2
XP_011524741.1
13
1845
XM_011526440.2
XP_011524742.1
13
1697
XM_017026265.1
XP_016881754.1
13
1744
XM_017026266.1
XP_016881755.1
13
1665
XM_017026267.1
XP_016881756.1
12
1515
XM_017026268.1
XP_016881757.1
11
1358
XM_017026269.1
XP_016881758.1
11
1468
XM_017026270.1
XP_016881759.1
11
1236
XM_017026271.1
XP_016881760.1
11
1325
XM_017026272.1
XP_016881761.1
12
1552
XM_017026273.1
XP_016881762.1
12
1537
GenBank mRNA
AB208884.1
-
7
732
AF357970.1
-
12
1577
AK074389.1
-
5
598
AK096516.1
-
14
1846
AK289713.1
-
13
1744
AK290092.1
-
13
1660
AK299866.1
-
10
1446
AK309087.1
-
12
1578
AL831876.1
-
3
250
BC029104.1
-
13
1619
3872727
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
carnitine metabolic process
carnitine O-palmitoyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0