Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136052.2 | NP_001129524.1 | 18 | 2382 | ||
NM_001199752.2 | NP_001186681.1 | 18 | 2479 | |||
NM_001199753.1 | NP_001186682.1 | 18 | 2415 | |||
NM_152359.2 | NP_689572.1 | 17 | 2346 | |||
NR_108072.1 | 19 | 2577 | ||||
XM_005258505.2 | XP_005258562.1 | 18 | 2452 | |||
XM_005258506.4 | XP_005258563.1 | 17 | 2368 | |||
XM_006723009.3 | XP_006723072.1 | 15 | 2184 | |||
XM_011526438.2 | XP_011524740.1 | 18 | 2508 | |||
XM_011526439.2 | XP_011524741.1 | 18 | 2572 | |||
XM_011526440.2 | XP_011524742.1 | 17 | 2289 | |||
XM_017026265.1 | XP_016881754.1 | 17 | 2336 | |||
XM_017026266.1 | XP_016881755.1 | 17 | 2257 | |||
XM_017026267.1 | XP_016881756.1 | 17 | 2242 | |||
XM_017026268.1 | XP_016881757.1 | 16 | 2085 | |||
XM_017026269.1 | XP_016881758.1 | 16 | 2195 | |||
XM_017026270.1 | XP_016881759.1 | 16 | 1963 | |||
XM_017026271.1 | XP_016881760.1 | 16 | 2052 | |||
XM_017026272.1 | XP_016881761.1 | 16 | 2144 | |||
XM_017026273.1 | XP_016881762.1 | 16 | 2129 | |||
GenBank mRNA | AF357970.1 | - | 17 | 2304 | ||
AK074389.1 | - | 10 | 1325 | |||
AK096516.1 | - | 19 | 2573 | |||
AK289713.1 | - | 18 | 2471 | |||
AK290092.1 | - | 18 | 2387 | |||
BC029104.1 | - | 18 | 2346 | 3872727 |