Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172689.1 | NP_001166160.1 | 6 | 1869 | ||
NM_001172690.1 | NP_001166161.1 | 5 | 1678 | |||
NM_001172691.1 | NP_001166162.1 | 5 | 1672 | |||
NM_001172692.1 | NP_001166163.1 | 4 | 1545 | |||
NM_152360.3 | NP_689573.3 | 8 | 1937 | |||
XM_011526446.2 | XP_011524748.1 | 6 | 1756 | |||
XM_017026275.1 | XP_016881764.1 | 6 | 1863 | |||
XM_017026276.1 | XP_016881765.1 | 6 | 2585 | |||
XM_017026277.1 | XP_016881766.1 | 6 | 1750 | |||
XM_017026278.1 | XP_016881767.1 | 4 | 1623 | |||
XM_017026279.1 | XP_016881768.1 | 4 | 1568 | |||
XM_017026280.1 | XP_016881769.1 | 5 | 1814 | |||
XM_017026281.1 | XP_016881770.1 | 4 | 2407 | |||
GenBank mRNA | AK055483.1 | - | 3 | 1687 | ||
AK074539.1 | - | 6 | 1865 | |||
BC015418.1 | - | 1 | 924 | |||
BC042170.2 | - | 4 | 1632 | 5014852 | ||
BC051263.1 | - | 1 | 499 | |||
BC064962.1 | - | 8 | 1907 | 6141400 |