Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319083.1 | NP_001306012.1 | 4 | 537 | ||
NM_173480.2 | NP_775751.1 | 4 | 636 | |||
XM_011527682.2 | XP_011525984.1 | 3 | 504 | |||
GenBank mRNA | AK302165.1 | - | 4 | 537 | ||
AK314848.1 | - | 4 | 558 | |||
BC028974.1 | - | 4 | 568 | 5138909 | ||
BX537601.1 | - | 4 | 577 | |||
EF534355.1 | - | 4 | 636 |