Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144617.2 | NP_653218.1 | 2 | 253 | ||
GenBank mRNA | AK056951.1 | - | 2 | 253 | ||
AK297852.1 | - | 2 | 248 | |||
BC068046.1 | - | 2 | 241 | 6074542 | ||
EU794622.1 | - | 2 | 253 |