Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173482.2 | NP_775753.2 | 7 | 1384 | ||
GenBank mRNA | AK097684.1 | - | 7 | 1384 | ||
AL833847.1 | - | 7 | 852 | |||
BC035678.1 | - | 7 | 1367 | 5742752 |