Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195833.1 | NP_001182762.1 | 7 | 586 | ||
NM_198445.3 | NP_940847.1 | 7 | 570 | |||
XM_006723015.2 | XP_006723078.1 | 7 | 580 | |||
XM_006723016.2 | XP_006723079.1 | 7 | 548 | |||
XM_011526454.2 | XP_011524756.1 | 7 | 650 | |||
XM_011526455.2 | XP_011524757.1 | 7 | 580 | |||
GenBank mRNA | AK126119.1 | - | 4 | 966 | ||
AK127808.1 | - | 7 | 545 | |||
AK298327.1 | - | 7 | 586 | |||
DA814564.1 | - | 7 | 546 |