Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256440.1 | NP_001243369.1 | 6 | 437 | ||
NM_001256441.1 | NP_001243370.1 | 6 | 432 | |||
NR_046202.1 | 6 | 485 | ||||
XM_005258520.2 | XP_005258577.1 | 6 | 429 | |||
XM_011526460.2 | XP_011524762.1 | 6 | 425 | |||
XM_017026291.1 | XP_016881780.1 | 6 | 440 | |||
XM_017026292.1 | XP_016881781.1 | 5 | 346 | |||
XM_017026293.1 | XP_016881782.1 | 6 | 440 | |||
XM_017026294.1 | XP_016881783.1 | 3 | 155 | |||
XM_017026295.1 | XP_016881784.1 | 7 | 535 | |||
GenBank mRNA | AK094207.1 | - | 6 | 425 | ||
AL834131.1 | - | 6 | 462 | |||
BC023623.2 | - | 6 | 463 | 4662735 | ||
BC027935.1 | - | 6 | 453 | 5202287 | ||
BI916290.1 | - | 6 | 432 | |||
BM459040.1 | - | 6 | 382 | |||
CR936760.1 | - | 6 | 438 |