Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256440.1 | NP_001243369.1 | 4 | 261 | ||
NM_001256441.1 | NP_001243370.1 | 4 | 256 | |||
NR_046202.1 | 4 | 309 | ||||
XM_005258520.2 | XP_005258577.1 | 4 | 253 | |||
XM_011526460.2 | XP_011524762.1 | 4 | 249 | |||
XM_017026291.1 | XP_016881780.1 | 4 | 264 | |||
XM_017026292.1 | XP_016881781.1 | 3 | 170 | |||
XM_017026293.1 | XP_016881782.1 | 4 | 264 | |||
XM_017026295.1 | XP_016881784.1 | 4 | 237 | |||
GenBank mRNA | AK094207.1 | - | 4 | 249 | ||
AL834131.1 | - | 4 | 286 | |||
BC023623.2 | - | 3 | 165 | 4662735 | ||
BC027935.1 | - | 4 | 277 | 5202287 | ||
BI916290.1 | - | 4 | 256 | |||
BM459040.1 | - | 4 | 206 | |||
CR936760.1 | - | 4 | 262 |