Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004948.3 | NP_004939.1 | 17 | 2849 | ||
NM_024421.2 | NP_077739.1 | 16 | 2803 | |||
GenBank mRNA | CU444724.1 | - | 2 | 181 | ||
X72925.1 | - | 17 | 2849 |