Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC163P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033296.1 | 5 | 863 | |||
NR_033297.1 | 5 | 906 | ||||
NR_033298.1 | 6 | 991 | ||||
GenBank mRNA | AK054714.1 | - | 5 | 875 | ||
AK300867.1 | - | 6 | 881 | |||
BC036618.2 | - | 5 | 524 | |||
BC047421.1 | - | 6 | 960 | |||
BC101254.2 | - | 5 | 801 | |||
BC101255.2 | - | 5 | 758 | |||
BC101256.1 | - | 5 | 758 | |||
BC101257.1 | - | 6 | 886 | |||
BC113060.1 | - | 6 | 814 | |||
BC113061.1 | - | 6 | 881 |