Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152367.2 | NP_689580.2 | 3 | 375 | ||
XM_011540623.2 | XP_011538925.1 | 3 | 507 | |||
XM_011540625.2 | XP_011538927.1 | 4 | 603 | |||
XM_011540626.2 | XP_011538928.1 | 3 | 429 | |||
XM_017000260.1 | XP_016855749.1 | 4 | 1840 | |||
GenBank mRNA | AK096035.1 | - | 3 | 375 | ||
BC128148.1 | - | 2 | 269 | 40112561 | ||
BC128149.1 | - | 2 | 269 | 40112562 |