Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162916.1 | NP_001156388.1 | 1 | 134 | ||
NM_138467.2 | NP_612476.1 | 1 | 130 | |||
NR_027962.1 | 1 | 108 | ||||
XM_006710347.1 | XP_006710410.1 | 1 | 143 | |||
GenBank mRNA | AK098237.1 | - | 1 | 130 | ||
AK294264.1 | - | 1 | 112 | |||
AK299849.1 | - | 1 | 85 | |||
AK311272.1 | - | 1 | 54 | |||
BC009514.1 | - | 1 | 64 | |||
BC071765.1 | - | 1 | 130 | 3931600 | ||
DA033034.1 | - | 1 | 134 |