Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182752.3 | NP_877429.2 | 3, 4 | 560 | ||
GenBank mRNA | AK025464.1 | - | 2, 3 | 280 | ||
AK096427.1 | - | 3, 4 | 537 | |||
AK289919.1 | - | 3, 4 | 510 | |||
AK298343.1 | - | 3, 4 | 399 |