Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195736.1 | NP_001182665.1 | 5, 6 | 651 | ||
NM_152371.3 | NP_689584.2 | 5, 6 | 597 | |||
NR_036637.1 | 5, 6 | 530 | ||||
XM_006710354.3 | XP_006710417.1 | 5, 6 | 585 | |||
XM_011540664.1 | XP_011538966.1 | 5, 6 | 641 | |||
GenBank mRNA | AF425266.1 | - | 5, 6 | 541 | ||
AK057027.1 | - | 5, 6 | 582 | |||
AK308034.1 | - | 5, 6 | 507 | |||
BC022547.1 | - | 5, 6 | 537 | 4812314 | ||
BQ709693.1 | - | 433 | ||||
DA495138.1 | - | 5, 6 | 530 |