Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 22 | 3002 | ||
GenBank mRNA | AK128233.1 | - | 14 | 2062 | ||
BC067101.1 | - | 22 | 3012 | 5928128 | ||
BX537911.1 | - | 21 | 2726 | |||
BX647453.1 | - | 22 | 2963 | |||
BX647455.1 | - | 22 | 3002 | |||
BX647644.1 | - | 22 | 2963 |