Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 33 | 3954 | ||
GenBank mRNA | AF086254.1 | - | 2 | 44 | ||
AK093280.1 | - | 6 | 840 | |||
AK128233.1 | - | 25 | 3014 | |||
BC014671.1 | - | 7 | 474 | |||
BX537911.1 | - | 32 | 3678 | |||
BX647453.1 | - | 33 | 3915 | |||
BX647455.1 | - | 33 | 3954 | |||
BX647644.1 | - | 33 | 3915 |