Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 10 | 2051 | ||
XM_005270457.3 | XP_005270514.1 | 11 | 2080 | |||
XM_011540677.2 | XP_011538979.1 | 6 | 1205 | |||
XM_017000294.1 | XP_016855783.1 | 10 | 2401 | |||
GenBank mRNA | AK296649.1 | - | 11 | 1946 | ||
AK299375.1 | - | 12 | 2136 | |||
AK300847.1 | - | 5 | 991 | |||
AK304599.1 | - | 6 | 1464 | |||
AK314096.1 | - | 10 | 1645 | |||
AY313607.1 | - | 10 | 1890 | |||
AY358294.1 | - | 12 | 2167 | |||
BC028292.1 | - | 10 | 1874 | 4938380 | ||
BC030608.2 | - | 10 | 1892 | 4826932 |