Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144622.2 | NP_653223.2 | 7 | 1112 | ||
XM_011509188.2 | XP_011507490.1 | 5 | 575 | |||
XM_011509189.2 | XP_011507491.1 | 2 | 964 | |||
GenBank mRNA | AK057496.1 | - | 7 | 1112 | ||
AK097468.1 | - | 5 | 526 | |||
AK097993.1 | - | 2 | 784 | |||
BC111794.1 | - | 7 | 1091 |