Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138795.3 | NP_620150.1 | 5 | 583 | ||
GenBank mRNA | AB118752.1 | - | 5 | 412 | ||
AK127138.1 | - | 5 | 536 | |||
AK313122.1 | - | 5 | 561 | |||
BC015408.1 | - | 5 | 514 | 4391821 | ||
BM548962.1 | - | 5 | 568 |