Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138796.3 | NP_620151.1 | 4 | 317 | ||
XM_005273052.1 | XP_005273109.1 | 4 | 299 | |||
XM_011509194.2 | XP_011507496.1 | 4 | 299 | |||
GenBank mRNA | AK098591.1 | - | 4 | 297 | ||
AK310291.1 | - | 4 | 301 | |||
AY963797.1 | - | 4 | 320 | |||
BC014608.2 | - | 4 | 288 | 4824429 |