Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138796.3 | NP_620151.1 | 8 | 859 | ||
XM_005273052.1 | XP_005273109.1 | 8 | 841 | |||
XM_006711165.3 | XP_006711228.1 | 7 | 799 | |||
GenBank mRNA | AK098591.1 | - | 8 | 839 | ||
AK310291.1 | - | 11 | 1181 | |||
AY963797.1 | - | 8 | 862 | |||
BC014608.2 | - | 8 | 830 | 4824429 |
Gene Symbol: SPATA17-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI025201.1 | - | 2 | 222 |