Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173484.3 | NP_775755.3 | 3 | 1079 | ||
XM_005270465.3 | XP_005270522.2 | 4 | 703 | |||
XM_011540700.1 | XP_011539002.1 | 6 | 1775 | |||
XM_011540704.2 | XP_011539006.1 | 4 | 646 | |||
GenBank mRNA | AK097479.1 | - | 3 | 1051 | ||
AK308458.1 | - | 5 | 1609 | |||
BC035408.1 | - | 3 | 1036 | |||
BC049844.1 | - | 3 | 1079 | 5166200 |