Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320244.1 | NP_001307173.1 | 4 | 1603 | ||
NM_144626.2 | NP_653227.1 | 4 | 1508 | |||
XM_005270467.3 | XP_005270524.1 | 3 | 1748 | |||
XM_011540705.2 | XP_011539007.1 | 4 | 1734 | |||
GenBank mRNA | BC016858.1 | - | 3 | 1361 | 3845811 | |
BC072393.1 | - | 4 | 1591 |