Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300834.1 | NP_001287763.1 | 6 | 491 | ||
NM_001300835.1 | NP_001287764.1 | 5 | 471 | |||
NM_174896.3 | NP_777556.1 | 6 | 566 | |||
XM_011540712.2 | XP_011539014.1 | 7 | 6046 | |||
XM_017000323.1 | XP_016855812.1 | 7 | 1840 | |||
XM_017000324.1 | XP_016855813.1 | 6 | 5951 | |||
GenBank mRNA | AK123160.1 | - | 3 | 1845 | ||
BC017973.1 | - | 6 | 453 | 4693393 | ||
BG545926.1 | - | 6 | 381 | |||
HX785231.1 | - | 5 | 353 |