Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001944.2 | NP_001935.2 | 11 | 1554 | ||
XM_011525850.1 | XP_011524152.1 | 11 | 1554 | |||
GenBank mRNA | AK290367.1 | - | 11 | 1554 | ||
M76482.1 | - | 11 | 1528 |