Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152864.3 | NP_690603.3 | 6 | 641 | ||
XM_011528527.1 | XP_011526829.1 | 6 | 603 | |||
XM_011528528.2 | XP_011526830.1 | 6 | 1065 | |||
XM_011528529.2 | XP_011526831.1 | 6 | 1051 | |||
XM_017027636.1 | XP_016883125.1 | 6 | 573 | |||
XM_017027637.1 | XP_016883126.1 | 6 | 1055 | |||
XM_017027638.1 | XP_016883127.1 | 6 | 1061 | |||
XM_017027639.1 | XP_016883128.1 | 6 | 1047 | |||
GenBank mRNA | AK126246.1 | - | 6 | 564 | ||
AK225765.1 | - | 6 | 595 | |||
BC041812.1 | - | 6 | 639 | 5302801 | ||
BG708360.1 | - | 6 | 644 |