Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178456.2 | NP_848551.1 | 1 | 84 | ||
GenBank mRNA | BC035405.1 | - | 1 | 73 | 5166377 | |
BX093883.1 | - | 1 | 84 |