Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008844.2 | NP_001008844.1 | 24 | 5113 | ||
NM_001319034.1 | NP_001305963.1 | 24 | 5581 | |||
NM_004415.3 | NP_004406.2 | 24 | 6910 | |||
GenBank mRNA | AB209992.1 | - | 24 | 5071 | ||
BC140802.1 | - | 24 | 5046 | 9021670 | ||
HM151899.1 | - | 24 | 5519 | |||
J05211.1 | - | 2 | 3212 | |||
M77830.3 | - | 24 | 6848 |