Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144628.3 | NP_653229.1 | 4, 5 | 675 | ||
NR_111901.1 | 4, 5 | 675 | ||||
XM_005260661.1 | XP_005260718.1 | 4, 5 | 665 | |||
XM_006723540.3 | XP_006723603.1 | 3, 4 | 454 | |||
XM_017027645.1 | XP_016883134.1 | 3, 4 | 446 | |||
GenBank mRNA | AB449906.1 | - | 4, 5 | 527 | ||
AK127062.1 | - | 4, 5 | 655 | |||
AK291648.1 | - | 4, 5 | 638 | |||
BC014983.1 | - | 4, 5 | 625 | 4861869 | ||
DA230581.1 | - | 1, 2 | 188 |